NanoDrop Ultra USB WiFi and Bluetooth Dongle
Thermo Scientific™
NanoDrop Ultra USB WiFi and Bluetooth Dongle
Purchase a NanoDrop™ Ultra USB WiFi and Bluetooth Dongle to enable Wi-Fi and Bluetooth connectivity to NanoDrop Ultra UV-Vis Spectrophotometers and Fluorometers.
Have Questions?
Catalog NumberIncludes
NDU-WIFI-STICKNanoDrop Ultra USB WiFi and Bluetooth Dongle
Catalog number NDU-WIFI-STICK
Price (INR)
11,632.00
Online offer
Ends: 31-Dec-2026
16,616.00
Save 4,984.00 (30%)
Each
Includes:
NanoDrop Ultra USB WiFi and Bluetooth Dongle
Price (INR)
11,632.00
Online offer
Ends: 31-Dec-2026
16,616.00
Save 4,984.00 (30%)
Each
Upgrade your wireless-enabled Thermo Scientific™ NanoDrop™ Ultra Spectrophotometer and Fluorometer with this high-power USB Wi-Fi and Bluetooth Dongle. Achieve high speed transfer rates and maximum range of connectivity during remote monitoring of your instrument.
Key features and benefits:
  • Leading wireless technology standard for top performance and range
  • Maximum speed ofup to 300 Mbps for fast wireless transfer rates
  • Extensive range with built-in high-power antenna
Specifications
For Use With (Equipment)NanoDrop Ultra Spectrophotometers and Fluorometer
IncludesNanoDrop Ultra USB WiFi and Bluetooth Dongle
Unit SizeEach