Neon™ NxT Electroporation System 8-Channel Pipette Station
Invitrogen™
Neon™ NxT Electroporation System 8-Channel Pipette Station
The Neon NxT Electroporation System 8-Channel Pipette Station is designed specifically for use with the Neon NxT Electroporation System.
Have Questions?
Catalog NumberQuantity
NEON8PS1 pipette station
Catalog number NEON8PS
Price (INR)
394,036.00
Each
Quantity:
1 pipette station
Price (INR)
394,036.00
Each
The Neon NxT Electroporation System 8-Channel Pipette Station is designed specifically for use with the Neon NxT Electroporation System (Cat. No. NEON18S). Each Neon NxT Electroporation System with 1-Channel and 8-Channel Pipettes (Cat. No. NEON18S and NEON18SK) comes with an 8-channel pipette station. Those looking to upgrade their Neon NxT Electroporation System with 1-Channel Pipette (Cat. No. NEON1S, NEON1SK) should pursue the Neon NxT Electroporation System 8-Channel Upgrade Package (Cat. No. NEON8 and NEON8U), which includes the Neon NxT Electroporation System 8-Channel Pipette, 8-channel pipette station, and 8-channel software activation. This pipette station is offered separately for those needing an additional 8-channel pipette station or replacement for their existing pipette station.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Transfection
For Use With (Equipment)Neon™ NxT Electroporation System
Quantity1 pipette station
Unit SizeEach