NetDose Pro Digital Dosimetry Solution
NetDose Pro Digital Dosimetry Solution
NetDose Pro Digital Dosimetry Solution
NetDose Pro Digital Dosimetry Solution
Thermo Scientific™
NetDose Pro Digital Dosimetry Solution
The Thermo Scientific™ NetDose™ Pro Digital Dosimetry Solution redefines the dose monitoring experience by delivering enhanced operational efficiency, cost reduction, and real-time access to dose records at both enterprise and user levels.
Have Questions?
Catalog NumberIP Rating
NETDOSEIP67
Catalog number NETDOSE
Price (INR)
-
IP Rating:
IP67
The Thermo Scientific™ NetDose™ Pro Digital Dosimetry Solution includes a wearable, connected device for monitoring radiation. The NetDose™ dosimeter redefines the dose monitoring experience by delivering enhanced operational efficiency, cost reduction, and real-time access to dose records at both enterprise and user levels. This NVLAP-accredited digital dosimeter (NVLAP Lab Code 600295-0) is tailored to adapt to your facility operations seamlessly.
The Thermo Scientific™ NetDose™ Pro Digital Dosimetry Solution includes a compact instrument designed to track and inform radiation exposure risk for personnel and help companies adhere to stringent regulatory requirements. The NetDose™ dosimeter does this by continuously measuring radiation levels and transmitting the data to either a personal mobile application or a web-based user interface, keeping both personal and customer information secure during the process.
NetDose™ represents a significant leap forward in personal dosimetry, replacing costly and labor-intensive methods that require sending devices back to manufacturers for analysis. The dosimeter uses Bluetooth 5.0 wireless technology to connect to a gateway data router and/or the NetDose™ mobile app, transmitting accurate and rapid data to an interface where users can monitor radiation levels and generate reports, as well as assign devices to wearers and manage their entire dosimetry system.
Each dosimeter can be individually programmed to transmit data at scheduled intervals, while the wearer can read radiation levels on-demand via a button found on the device. Importantly, the entire system – the NetDose dosimeter, gateway router, mobile app and user interface – is secure, and the NetDose only stores measured radiation dose data in its memory, omitting customer and wearer information. The device is perfect for healthcare professionals working near radiation sources – such as radiologists – as well as any personnel frequently in proximity to high energy radiation, including industrial scrap metal recyclers, veterinarians and dentists.
  • Real-time insights: The NetDose offers accurate and rapid readings with a low limit of detection. On-demand and preprogrammed radiation dose readings offer immediate insights without removing the device, informing immediate action to reduce radiation exposure risk.
  • Reliable performance: With a long life and replaceable battery, NetDose is also stable over a wide temperature range (-20 to 40 °C), making it appropriate for a variety of monitoring environments.
  • Ease of Use: Customizable user interface generates personalized reports and graphs, as well as tailored reading intervals. Compact and lightweight (~30 g) design makes it comfortable for the wearer, while the connected platform eliminates the need to exchange the device with the manufacturer or navigate badge reassignment challenges.
Specifications
DescriptionNetDose Pro Digital Dosimetry Solution
IP RatingIP67
Operating Temperature (Metric) -20°C to 40°C
Unit SizeEach