NuPAGE™ MES SDS Running Buffer (Powder)
Invitrogen™

NuPAGE™ MES SDS Running Buffer (Powder)

Invitrogen NuPAGE MES SDS Running Buffer (Powder) is formulated for use with Bis-Tris gels to deliver fast SDS-PAGE separations of small- to medium-sized proteins. Supplied as a powder for flexible preparation, room temperature storage, and a long 2-year shelf life.
Have Questions?
Catalog NumberQuantity
NP0001055 Packets
Catalog number NP000105
Price (INR)
4,352.00
Each
Quantity:
5 Packets
Price (INR)
4,352.00
Each

Invitrogen NuPAGE MES SDS Running Buffer (Powder) is formulated for use with Bis-Tris gels to enable enfast, high-resolution SDS-PAGE protein separations. The MES buffer system supports rapid protein migration and is recommended for resolving small- to medium-sized proteins. Supplied as a dry powder, this running buffer enables flexible preparation, room temperature storage, a two-year shelf life, and is designed to be a convenient and reliable choice for routine electrophoresis workflows.

Features and benefits include:

  • Fast separations for small- to medium-sized proteins—MES supports faster protein migration than MOPS-based buffers, helping reduce run times while maintaining resolution
  • Optimized for Bis-Tris gel workflows—designed for use with Bis-Tris gels including both NuPAGE Bis-Tris and Bolt Bis-Tris Plus gels
  • Powder format for flexible preparation—prepare fresh running buffer as needed to support consistent electrophoresis performance and simplify storage
  • Room temperature storage and extended shelf life—powder format supports convenient storage with a two-year shelf life for inventory efficiency
  • Reliable performance for routine SDS-PAGE—supports consistent results across common protein electrophoresis workflows when used under recommended conditions
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)SDS-PAGE
For Use With (Equipment)Electrophoresis Chambers
FormulationMES SDS Running Buffer
Quantity5 Packets
Shelf Life2 Years
Storage RequirementsRoom Temperature
Sufficient ForPreparing 5 L Running Buffer
ColorWhite
FormatPowder
Gel TypeBis-Tris
Product LineNuPAGE
Product TypeSDS Running Buffer
Unit SizeEach