NuPAGE™ Bis-Tris XCell SureLock™ Mini-Cell Welcome Pack, 4 to 12%
Thermo Scientific™

NuPAGE™ Bis-Tris XCell SureLock™ Mini-Cell Welcome Pack, 4 to 12%

The NuPAGE Bis-Tris Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWells
NP032D10-well
NP032E12-well
NP032F15-well
Catalog number NP032D
Price (INR)
58,573.00
キャンペーン価格
Ends: 31-Dec-2026
83,675.00
Save 25,102.00 (30%)
Each 1
Wells:
10-well
Price (INR)
58,573.00
キャンペーン価格
Ends: 31-Dec-2026
83,675.00
Save 25,102.00 (30%)
Each 1
The NuPAGE Bis-Tris Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell. The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels.

The NuPAGE Bis-Tris Welcome Pack contains:

  • XCell SureLock Mini-Cell (EI0001)
  • NuPAGE Bis-Tris mini gels (2 boxes, 20 gels)
  • NuPAGE MES SDS Running Buffer, 20X (NP0002)
  • MES and MOPS Running Buffer, Powder (NP000105, NP000205)
  • NuPAGE LDS Sample Buffer, 4X (NP0007)
  • NuPAGE Sample Reducing Agent, 10X (NP0009)
  • PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)

About the XCell SureLock Mini-Cell

The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels. The Mini-Cell is simple, sturdy, and easy to use. Each XCell SureLock Mini-Cell can accommodate up to two gels per run. Run times may vary depending on buffer conditions and power supplies used.

About NuPAGE Bis-Tris gels

Invitrogen NuPAGE Bis-Tris Protein gels are precast polyacrylamide gels designed to give optimal separation of a wide range of proteins under denaturing conditions. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is crucial. Also, use NuPAGE gels for optimal results during day-to-day use.

Transfer

For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer Buffer (NP0006) for traditional wet transfer using the XCell II Blot Module (EI9051). Alternatively, rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)XCell SureLock Mini-Cell
Gel Thickness1.0 mm
Quantity1 Welcome Pk.
Shipping ConditionApproved for shipment at room temperature and wet ice
Gel Percentage4 to 12%
Gel SizeMini
Gel TypeBis-Tris
Product LineNuPAGE
Separation Range3.5 to 260 kDa
Wells10-well
Unit SizeEach 1