Tetra Core Set and NuPAGE™ Bis-Tris 4-12% Gels Combos
Invitrogen™

Tetra Core Set and NuPAGE™ Bis-Tris 4-12% Gels Combos

The Tetra Electrophoresis Core Set and NuPAGE Bis-Tris 4-12% Combo includes the Tetra Electrophoresis primary and companion cores along with three boxes of NuPAGE Bis-Tris gels. Easily swap and run up to four NuPAGE gels in Bio-Rad's Mini-PROTEAN™ Tetra Cell tank, achieving superior protein resolution and reproducibility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWells
NP032TS-10W10-well
NP032TS-12W12-well
NP032TS-15W15-well
Catalog number NP032TS-10W
Price (INR)
52,454.00
Each
Wells:
10-well
Price (INR)
52,454.00
Each

The Invitrogen Tetra Electrophoresis Core Set and NuPAGE Bis-Tris 4-12% Combo provides a comprehensive solution for running high-performance Invitrogen precast protein gels in Bio-Rad's Mini-PROTEAN Tetra Cell. This combo product includes the Tetra Electrophoresis primary and companion cores along with three boxes of NuPAGE Bis-Tris gels, allowing for seamless integration and efficient protein analysis applications such as western blotting and SDS-PAGE. Its intuitive design ensures ease of use, making it accessible for both new and experienced users.

Contents of the combo kit

  • Tetra Electrophoresis Primary Core (Cat. No. 1658037)
  • Tetra Electrophoresis Companion Core (Cat. No. 1658038)
  • 3 boxes of NuPAGE Bis-Tris 4-12% gels (10 gels per box)

About the Tetra Electrophoresis Core Set

The Tetra Electrophoresis cores stand out due to their compatibility with all Invitrogen mini gels, allowing users to swap and run gels effortlessly while achieving high-performance results. When using both the primary and companion cores, up to four gels can be run simultaneously, which significantly increases throughput and efficiency. Additionally, the self-aligning cassettes and leak-proof design ensure stable and reliable electrophoresis runs, making experiment setup easy and fast.

The Tetra Electrophoresis Primary Core comes with the Tetra Buffer Dam (Cat. No. 1658039) for use when running only one gel. To run three or four gels at once, the Tetra Electrophoresis Companion Core is used alongside the Primary Core. This combination allows for the simultaneous running of multiple gels, further enhancing throughput and efficiency in your lab. The Companion Core works seamlessly with the Primary Core, providing a flexible solution for varying experimental needs.

This cost-effective solution eliminates the need for new electrophoresis equipment, saving costs while enhancing your lab's capabilities. The Tetra Electrophoresis Core Set is straightforward to use: simply swap the cores in your Bio-Rad Mini-PROTEAN Tetra system for the Invitrogen Tetra cores and use Invitrogen mini protein gels following their recommended protocols.

About NuPAGE Bis-Tris gels

Invitrogen NuPAGE Bis-Tris Protein gels are precast polyacrylamide gels designed to give optimal separation of a wide range of proteins under denaturing conditions. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and other techniques where protein integrity is crucial. Also, use NuPAGE gels for optimal results during day-to-day use. Each box contains 10 gels.

For Research Use Only.
Specifications
AdaptersTetra Electrophoresis Cores
BufferNuPAGE MES SDS Running Buffer, NuPAGE MOPS SDS Running Buffer
For Use With (Equipment)Bio-Rad Mini Protean Tanks
Gel Thickness1.0 mm
Length (Metric)8 cm
Mode of SeparationMolecular Weight
QuantityTetra Core Set, 3 boxes of gels (30 gels)
Sample Loading VolumeUp to 60 μL
Shelf Life16 Months
Shipping ConditionRoom Temperature
Width (Metric)8 cm
Gel Percentage4 to 12%
Gel SizeMini
Gel TypeBis-Tris
Product LineNuPAGE
Product TypeElectrophoresis Cores and Precast Protein Gels
Separation Range3.5 to 260 kDa
Separation TypeDenaturing
Separation ofProtein
Wells10-well
Unit SizeEach
Contents & Storage
Store at 4°C to 25°C. Do not freeze.