Neon™ NxT Electroporation System 1-Channel Tubes
Invitrogen™
Neon™ NxT Electroporation System 1-Channel Tubes
Neon NxT Electroporation System 1-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 1-Channel Pipette.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
NT2424 Tubes
NT9696 tubes
Catalog number NT24
Price (INR)
20,440.00
Each
Quantity:
24 Tubes
Price (INR)
20,440.00
Each
Neon NxT Electroporation System 1-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 1-Channel Pipette (Cat. Nos. NEON1S, NEON1SK, NEON18S, NEON18SK). Each Neon NxT Electroporation System Kit for 25 x 2 reactions (Cat. Nos. N1025 and N10025) comes with eight 1-channel tubes, and each Neon NxT Electroporation System Kit with 1-Channel Tubes for 96 x 2 reactions (Cat. Nos. N1096 and N10096) comes with 32 1-channel tubes. This pack of additional tubes is offered for those wishing to use single tubes per reaction depending on their experimental setup or for reactions with the 1-channel pipette when predominately using the 8-channel pipette and utilizing kits that contain the 8-channel tubes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
IncludesIncludes 24 tubes, use at room temperature under normal laboratory conditions. Refer to the Neon™ NxT Electroporation System user manual for protocol instructions.
Quantity24 Tubes
For Use With (Application)Electroporation
For Use With (Equipment)Neon NxT Electroporation System
Unit SizeEach
Frequently asked questions (FAQs)
How are the Neon NxT Electroporation System 1-Channel Tubes (Cat. No. NT24 and NT96) packaged?
The tubes are packaged in groups of 4. Therefore, Cat. No. NT24 will have 6 bags of 4 tubes and Cat. No. NT96 will have 24 bags of 4 tubes.
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.