Neon™ NxT Electroporation System 8-Channel Tubes
Neon™ NxT Electroporation System 8-Channel Tubes
Neon™ NxT Electroporation System 8-Channel Tubes
Invitrogen™
Neon™ NxT Electroporation System 8-Channel Tubes
Neon NxT Electroporation System 8-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 8-Channel Pipette.
Have Questions?
Catalog NumberQuantity
NT88 Tubes
Catalog number NT8
Price (INR)
29,353.00
線上優惠
Ends: 30-Sep-2026
53,369.00
Save 24,016.00 (45%)
Each
Quantity:
8 Tubes
Price (INR)
29,353.00
線上優惠
Ends: 30-Sep-2026
53,369.00
Save 24,016.00 (45%)
Each
Neon NxT Electroporation System 8-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 8-Channel Pipette (Cat. Nos. NEON18S, NEON18SK, NEON8, NEON8U). Each Neon NxT Electroporation System Kit with 8-Channel Tubes for 96 x 2 reactions (Cat. Nos. N1096-8 and N10096-8) comes with four 8-channel tubes and each Neon NxT Electroporation System Kit for 384 x 2 reactions (Cat. Nos. N10384-8 and N100384-8) comes with sixteen 8-channel tubes. This pack of eight tubes is offered for those wishing to use one tube per eight reactions (tubes can be used up to twelve times) depending on their experimental setup.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
IncludesIncludes 8 x 8-chamber tubes, use at room temperature under normal laboratory conditions. Refer to the Neon™ NxT Electroporation System user manual for protocol instructions.
Quantity8 Tubes
For Use With (Application)Electroporation
For Use With (Equipment)Neon NxT Electroporation System
Unit SizeEach