Tetra Core Set and Bolt™ Bis-Tris Plus 4-12% Gels Combos
Tetra Core Set and Bolt™ Bis-Tris Plus 4-12% Gels Combos
Invitrogen™

Tetra Core Set and Bolt™ Bis-Tris Plus 4-12% Gels Combos

The Tetra Electrophoresis Core Set and Bolt Bis-Tris Plus 4-12% Combo includes the Tetra Electrophoresis primary and companion cores along with three boxes of Bolt Bis-Tris Plus gels. Easily swap and run up to four Bolt Bis-Tris gels in Bio-Rad's Mini-PROTEAN™ Tetra Cell tank with a run time as short as 20 minutes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWells
NW0412TS-12W12-well
NW0412TS-10W10-well
NW0412TS-15W15-well
Catalog number NW0412TS-12W
Price (INR)
34,139.00
Online offer
Ends: 31-Mar-2026
56,898.00
Save 22,759.00 (40%)
Each
Add to cart
Wells:
12-well
Price (INR)
34,139.00
Online offer
Ends: 31-Mar-2026
56,898.00
Save 22,759.00 (40%)
Each
Add to cart

The Invitrogen Tetra Electrophoresis Core Set and Bolt Bis-Tris Plus 4-12% Combo provides a comprehensive solution for running high-performance Invitrogen precast protein gels in Bio-Rad's Mini-PROTEAN Tetra Cell. This combo product includes the Tetra Electrophoresis primary and companion cores along with three boxes of Bolt Bis-Tris Plus gels, allowing for seamless integration and efficient protein analysis applications such as western blotting and SDS-PAGE. Its intuitive design ensures ease of use, making it accessible for both new and experienced users.

Contents of the combo kit

  • Tetra Electrophoresis Primary Core (Cat. No. 1658037)
  • Tetra Electrophoresis Companion Core (Cat. No. 1658038)
  • 3 boxes of Bolt Bis-Tris Plus 4-12% gels (10 gels per box)

About the Tetra Electrophoresis Core Set

The Tetra Electrophoresis cores stand out due to their compatibility with all Invitrogen mini gels, allowing users to swap and run gels effortlessly while achieving high-performance results. When using both the primary and companion cores, up to four gels can be run simultaneously, which significantly increases throughput and efficiency. Additionally, the self-aligning cassettes and leak-proof design ensure stable and reliable electrophoresis runs, making experiment setup easy and fast.

The Tetra Electrophoresis Primary Core comes with the Tetra Buffer Dam (Cat. No. 1658039) for use when running only one gel. To run three or four gels at once, the Tetra Electrophoresis Companion Core is used alongside the Primary Core. This combination allows for the simultaneous running of multiple gels, further enhancing throughput and efficiency in your lab. The Companion Core works seamlessly with the Primary Core, providing a flexible solution for varying experimental needs.

This cost-effective solution eliminates the need for new electrophoresis equipment, saving costs while enhancing your lab's capabilities. The Tetra Electrophoresis Core Set is straightforward to use: simply swap the cores in your Bio-Rad Mini-PROTEAN Tetra system for the Invitrogen Tetra cores and use Invitrogen mini protein gels following their recommended protocols.

About Bolt Bis-Tris Plus Gels

Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal broad range separation of proteins under denaturing conditions. Similar to NuPAGE Bis-Tris gels, Bolt Bis-Tris Plus gels offer excellent protein integrity with neutral pH, a wide range of molecular weight separation, fast run times of 20–35 minutes, and a long shelf life of up to 16 months at room temperature. Bolt Bis-Tris Plus gels also have the unique WedgeWell design that allows for easier sample loading and up to 2X more sample volume than other standard precast gels. Bolt Bis-Tris Plus gels come in four percentages and multiple well formats. Each box contains 10 gels.

For Research Use Only.
Specifications
AdaptersTetra Electrophoresis Cores
BufferBolt MES SDS Running Buffer, Bolt MOPS SDS Running Buffer
For Use With (Equipment)Bio-Rad Mini Protean Tanks
Gel Thickness1.0 mm
Length (Metric)8 cm
Mode of SeparationMolecular Weight
QuantityTetra Core Set, 3 boxes of gels (30 gels)
Sample Loading VolumeUp to 45 μL
Shelf Life16 Months
Shipping ConditionRoom Temperature
Width (Metric)8 cm
Gel Percentage4 to 12%
Gel SizeMini
Gel TypeBis-Tris Plus
Product LineBolt, WedgeWell
Product TypeElectrophoresis Cores and Precast Protein Gels
Separation Range3.5 to 260 kDa
Separation TypeDenaturing
Separation ofProtein
Wells12-well
Unit SizeEach
Contents & Storage
Store at 4°C to 25°C. Do not freeze.