Olympus™ 2X Objective, PlanApo, 0.08 NA
Thermo Scientific™
Olympus™ 2X Objective, PlanApo, 0.08 NA
Expand the functionality of your Thermo Scientific™ high-content instrument
Have Questions?
Catalog NumberQuantity
NX1002X1 Ea.
Catalog number NX1002X
Price (INR)
450,674.00
Online offer
Ends: 30-Sep-2026
751,123.06
Save 300,449.06 (40%)
Each
Quantity:
1 Ea.
Price (INR)
450,674.00
Online offer
Ends: 30-Sep-2026
751,123.06
Save 300,449.06 (40%)
Each
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
The Olympus 2X (0.08NA) PlanApo objective lens is suitable for use with the Thermo Scientific CellInsight CX5, CX7 LED, and CX7 LZR high content screening (HCS) platforms. It has a magnification power suited for subcellular imaging and a numerical aperture that strikes a balance between resolution and versatility.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionOlympus™ 2X Objective, PlanApo (0.08 NA)
For Use With (Equipment)CellInsight CX5 HCS Platform
Lens TypeDry
Magnification2x (0.08NA)
Quantity1 Ea.
Product LineOlympus
TypeObjective Lens
Unit SizeEach