Olympus™ 4X Objective, U Plan S-Apo, 0.16 NA
Thermo Scientific™
Olympus™ 4X Objective, U Plan S-Apo, 0.16 NA
The Olympus 4X (0.16NA) U Plan S-Apo objective lens is suitable for use with the Thermo Scientific CellInsight CX5, CX7Read more
Have Questions?
Catalog NumberQuantity
NX1004X1 each
Catalog number NX1004X
Price (INR)
764,075.25
Each
Quantity:
1 each
Price (INR)
764,075.25
Each
The Olympus 4X (0.16NA) U Plan S-Apo objective lens is suitable for use with the Thermo Scientific CellInsight CX5, CX7 LED, and CX7 LZR high content screening (HCS) platforms. It has a magnification power suited for subcellular imaging and a numerical aperture which strikes a balance between resolution and versatility.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionOlympus™ 4X Objective, U Plan S-Apo (0.16 NA)
For Use With (Equipment)CellInsight CX5 HCS Platform
Lens TypeDry
Magnification4x (0.16NA)
Quantity1 each
Product LineOlympus
TypeObjective Lens
Unit SizeEach