1-Octane Sulfonic Acid Sodium Salt, Ion Pair Chromatography, Fisher Chemical™
1-Octane Sulfonic Acid Sodium Salt, Ion Pair Chromatography, Fisher Chemical™
1-Octane Sulfonic Acid Sodium Salt, Ion Pair Chromatography, Fisher Chemical™
Fisher Chemicals

1-Octane Sulfonic Acid Sodium Salt, Ion Pair Chromatography, Fisher Chemical™

C8H17NaO3S, CAS Number-5324-84-5, 1-octanesulfonate, sodium, sodium octylsulfonate, ipc-alks-8, 1-octanesulfonic acid, sodium salt, 1-octanesulfonic acid sodium salt, octyl sodium sulfonate, sodium octanesulfonate, sodium octane-1-sulfonate, sodium 1-octanesulfonate, unii-j46ez5jyb3, 100g, White
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
View all
Plastic powder jar
Catalog number O002846
also known as O/0028/46
HSN 29041010
Price (INR)
7,239.00
Each
Quantity:
25 g
Packaging:
Plastic powder jar
Request bulk or custom format
Price (INR)
7,239.00
Each
Specifications
Melting Point300°C
CAS5324-84-5
ColorWhite
pH5.5 to 7.5
Physical FormPowder/Solid
View more
C8H17NaO3S, CAS Number-5324-84-5, 1-octanesulfonate, sodium, sodium octylsulfonate, ipc-alks-8, 1-octanesulfonic acid, sodium salt, 1-octanesulfonic acid sodium salt, octyl sodium sulfonate, sodium octanesulfonate, sodium octane-1-sulfonate, sodium 1-octanesulfonate, unii-j46ez5jyb3, 100g, White