MAS™ OmniCORE II Control
Thermo Scientific™

MAS™ OmniCORE II Control

OmniCORE II controls consolidate general chemistry and serum protein QC processing into a single product offering three distinct levels with Thermo ScientificTM MASTM OmniCORE II Controls.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLevel
OCRL-2022
OCRL-1011
OCRL-3033
Catalog number OCRL-202
Price (INR)
-
Level:
2
Consolidate general chemistry and serum protein QC processing into a single product offering three distinct levels with Thermo Scientific MAS Omni·CORE II Controls. Value assignment is provided for key integrated instrument systems.
  • Three distinct levels
  • Liquid, ready-to-use
  • Human-serum based matrix
  • Assayed values available for most analyzers
Available in select countries only
Specifications
DisposableReusable
For Use With (Equipment)Key integrated instrument systems.
FormatVial
Level2
Quantity6x5 mL
Shelf Life36 Months at -25°C to -15°C
Stability30 days open-vial at 2°C to 8°C
TypeThird-party Quality Control
Detectable AnalytesAcetaminophen, Albumin, Alkaline Phosphatase, alpha-1-Acid Glycoprotein, alpha-1-Antitrypsin, alpha-2-Macroglobulin, ALT, Amikacin, Amylase, Amylase (Pancreatic), Antistreptolysin O (ASO), Apolipoprotein A1, Apolipoprotein B, AST, beta-2-microglobulin (B2M), Bile Acids, Bilirubin Conjugated, Bilirubin Direct (DBIL), Bilirubin Neonatal, Bilirubin Total (TBIL), Bilirubin Unconjugated, BUN, C3 Complement, C4 Complement, Caffeine, Calcium, Carbamazepine, Ceruloplasmin, Chloride, Cholesterol, Creatine Kinase (CK), CO2, Copper, Cortisol, C-Reactive Protein (CRP), Digoxin, Disopyramide, Ethanol, Ethosuximide, Ferritin, Gentamicin, GGT, Glucose, Glutamate Dehydrogenase (GLDH), Haptoglobin, HDL, IgA, IgE, IgG, IgM, Iron, Iron Binding Capacity (IBC), Iron Binding Capacity, Total (TIBC), Iron Binding Capacity (IBC), Iron Binding Capacity, Unsaturated (UIBC), Lactic Acid, LDH, LDL, Lidocaine, Lipase, Lipoprotein (a), Lithium, Magnesium, Methotrexate, N-acetylprocainamide (NAPA), Osmolality, Phenobarbital, Phenytoin, Phospholipids, Phosphorus, Potassium, Prealbumin, Primidone, Procainamide, Protein Electrophoresis, Pseudocholinesterase (PCHE), Quinidine, Rheumatoid Factor, Salicylate, Sodium, Thyroxine (T4 Total), Theophylline, Tobramycin, Total Protein, Transferrin (TRF), Tricyclic Antidepressants, Triglycerides, Thyroid Stimulating Hormone (TSH), T-uptake, Uric Acid, Valproic Acid, Vancomycin, Zinc
FormLiquid
Sample TypeSerum