TraceFinder™ Software
TraceFinder™ Software
Thermo Scientific™
TraceFinder™ Software
Routine quantitation and screening isn't about tedious method development, peak integration or laborious data review: go from sample to results with speed and confidence.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSoftware
OPTON-310045.1 SP1 Upgrade 3.3 Series
OPTON-310015.1 SP1
OPTON-310025.1 SP1 General Upgrade
OPTON-310035.1 SP1 Upgrade 4 Series
OPTON-310055.1 SP1 Upgrade 3 Series
OPTON-310065.1 SP1 Upgrade 5.0 Series
OPTON-310075.1 SP1 (Demo License)
OPTON-310375.1 SP1 (5+ Licenses)
Catalog number OPTON-31004
Price (INR)
-
Request A Quote
Software:
5.1 SP1 Upgrade 3.3 Series
Routine high-throughput screening and quantitation demands rapid access to confident results where users may have different experience levels.
Increase your productivity through acquiring and processing your targeted screening and routine quantitative data from LC and GC (including multiplexed and third-party systems), Quadrupole and HRAM-MS systems.
Configure with environmental, clinical, food safety, forensics and pharma/biopharma specific nomenclature, to rapidly develop confident methods which are accessible to all with minimal training requirements.
When used with Thermo Scientific™ High-Resolution Accurate Mass or Quadrupole mass spectrometry systems, TraceFinder software offers the most comprehensive quantitative and qualitative workflows to address critical challenges in your routine or advanced assays.
Discover the Unique Benefits of TraceFinder
  • Complete configurability for your application, and experience level
  • Quantify more in less time with pre-configured methods, reports and compound databases
  • Access the latest mass spectral libraries and compound databases for screening, or to provide additional confirmation
  • Easily manage your data across a network
Meets Application-Specific Requirements
  • The flexibility to configure TraceFinder software for general quantitative and qualitative workflows, or for specific applications such as environmental, clinical, food safety, forensics and pharma/biopharma
Increases Productivity
  • Simplifies workflow-driven method setup with fully automated data acquisition, data processing, and reporting
  • Automated decision making based upon user-defined parameters for more right-first-time analyses
  • Speeds data processing and reporting with compound database (CDB), smart calibration curves, library search capability, and custom reporting
  • Facilitates data review with multi-peak review capabilities, automatic retention time and ion ratio adjustment, and extensive flagging options
  • Simplifies operation with system start-up and shut-down methods
  • Provides quantitative, semi-quantitative, and screening workflows
Specifications, terms, and pricing are subject to change. Not all products are available in all countries. Please consult your local sales representative for details.
Specifications
For Use With (Equipment)Mass Spectrometers
Software5.1 SP1 Upgrade 3.3 Series
TypeSoftware Upgrade
Unit SizeEach