Isopropanol, GMP, Multi-Compendial
Isopropanol, GMP, Multi-Compendial
Greenfield Global

Isopropanol, GMP, Multi-Compendial

This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or aRead more
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 1 gal.
55 gal.
Catalog number P211USPNFDM55M
Price (INR)
-
Request A Quote
Quantity:
55 gal.
This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes. It is not intended for use as an active ingredient in drug manufacturing, nor as a medical device or disinfectant. Appropriate/legal use of this product is the responsibility of the user.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point83°C
CAS67-63-0
Chemical Name or MaterialIsopropyl Alcohol
ColorColorless
Molecular FormulaC3H8O
PackagingMetal drum
SynonymIsopropanol, Isopropyl Alcohol, IPA
GradeUSP
Molecular Weight (g/mol)18.02
Percent Purity68.0–72.0%
Quantity55 gal.
Unit Size55 gal.