Acetonitrile, ACS
Greenfield Global

Acetonitrile, ACS

This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
Have Questions?
Catalog number P300000ACSPL05M
Price (INR)
-
This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes. It is not intended for use as an active ingredient in drug manufacturing, nor as a medical device or disinfectant. Appropriate/legal use of this product is the responsibility of the user.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point178.9°F (81.6°C) at 101.325 kPa
CAS75-05-8
Chemical Name or MaterialAcetonitrile
ColorClear
Molecular FormulaC2H3N
PackagingCase
SynonymEthanenitrile, Methyl Cyanide, Cyanomethane
GradeUSP
Molecular Weight (g/mol)41.05
Percent Purity100%
Quantity5 gal.
Unit Size5 gal.