Benzyl Alcohol, GMP, Multi-Compendial
Greenfield Global

Benzyl Alcohol, GMP, Multi-Compendial

This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
Have Questions?
Catalog number P303WORLDCS4L
Price (INR)
-
This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
Not intended for Parenteral Use. This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes. It is not intended for use as an active ingredient in drug manufacturing, nor as a medical device or disinfectant. Appropriate/legal use of this product is the responsibility of the user.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
ColorColorless
PackagingCase
Boiling Point205°C
GradeMulti-Compendial
Molecular Weight (g/mol)108.14
Percent Purity98.0–100.5%
Physical FormLiquid
Quantity55 gal.
Unit Size55 gal.