pHrodo™ Dyes for Amine Labeling
Invitrogen™

pHrodo™ Dyes for Amine Labeling

Amine-reactive and TFP ester pHrodo dyes enable accurate measurement of cell endocytosis and phagocytosis, with low signal-to-background, for use in live cell assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorExcitation/EmissionReactive MoietyQuantity
P36013Green505/525Active Ester, STP Ester3 x 100 μg
P36011Red560/585Active Ester, STP Ester3 x 100 μg
P36010Red560/585Active Ester, STP Ester1 mg
P36012Green505/525Active Ester, STP Ester1 mg
P35358Deep Red640/655TFP ester3 x 100 μg
P35359Deep Red640/655TFP ester1 mg
Catalog number P36013
Price (INR)
31,579.00
線上優惠
Ends: 30-Sep-2026
52,631.00
Save 21,052.00 (40%)
Each
Color:
Green
Excitation/Emission:
505/525
Reactive Moiety:
Active Ester, STP Ester
Quantity:
3 x 100 μg
Price (INR)
31,579.00
線上優惠
Ends: 30-Sep-2026
52,631.00
Save 21,052.00 (40%)
Each
Perform cell internalization studies with low signal-to-background and high specificity using the pHrodo pH sensor dyes, including pHrodo Red, Green, and Deep Red dyes. These dyes can be conjugated to biomolecules such as antibodies, and become highly fluorescent only when found in acidic environments, such as those of cell lysosomes and endosomes. This enables detection of the conjugated biomolecule in the live cell assay without the need for wash steps or quencher dyes in the experimental workflow.
pHrodo Red, Green, and Deep Red dyes
The amine-reactive pH-sensitive pHrodo Red, Green, and Deep Red dyes are optimized for the creation of bioconjugates used in the study of antibody internalization, endocytosis, and phagocytosis in live cell assays. Amine-reactive pHrodo dyes enable you to label primary amines on an antibody, protein, cell, virus, or other amine-containing biomolecule to create a conjugate that fluoresces inside acidic environments but shows little to no signal at neutral pH. The lack of fluorescence of pHrodo dyes in a typical extracellular environment eliminates the need for wash steps or quencher dyes in the experimental workflow.

pHrodo iFL Red and Green STP Ester dyes
pHrodo iFL Red and Green STP Ester dyes are improved versions of the original pHrodo Red and Green dyes and are optimized to create more soluble conjugates than the original dyes. This enhanced property makes them useful for the labeling of antibodies that may otherwise precipitate out of solution during conjugation.

pHrodo Deep Red TFP Ester
The pHrodo Deep Red TFP Ester is distinct from pHrodo iFL Red and Green Ester dyes in that it has a lower pKa and, as a result, shows both improved signal-to-background ratios and fluoresces later in the endocytic pathway. Our low-background pHrodo Deep Red dye makes it easier than ever to study internalization with a higher certainty of results and less optimization because pHrodo Deep Red dye only fluoresces in the late endosome and lysosome.

Features and benefits of the pHrodo dyes for amine labeling

Customization—use pH-sensitive pHrodo dyes to make pH-sensitive bioconjugates of your choice
Speed and accuracy—get faster and more accurate results than with any other endocytosis or phagocytosis assay, no need for wash steps or quenchers
Multiplexing—multiplex with green or red fluorescent markers such as GFP or RFP with three different color choices
Superior signal window—with pHrodo Deep Red dye’s minimal background at neutral pH, maximal signal-to-noise ratio is enabled for certainty of results
Low pKa—pHrodo Deep Red dye eliminates non-specific signal from non-internalized cargo, resulting in fluorescence only in late endosomes and lysosomes

The increase in fluorescence of pHrodo dyes as the pH acidifies correlates with the acidification of intracellular vesicles, making these dyes valuable tools for the study of endocytosis or phagocytosis and their regulation by environmental factors, drugs, or pathogens. The spectral properties of pHrodo dyes also make them useful for multi-color experiments. pHrodo iFl Red, iFL Green, and Deep Red dyes can be detected using most standard red, green, and deep red fluorescent filter sets. pHrodo dye conjugates have been validated for use in a variety of applications, including flow cytometry, fluorescent microscopy, and high content screening (HCS).

pHrodo Deep Red dye has a lower pKa by design and shows no signal in neutral conditions, fluorescing only in acidic environments. This unique property enables rapid assay development and certainty of results when investigating antibody internalization and endocytic/phagocytic pathways. pHrodo Deep Red dye enables better discrimination of internalized cargo from outside the cell, whereas pHrodo iFl Red and Green dyes fluoresce in the early endosome.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
DescriptionpHrodo iFL Green STP ester, amine reactive dye
Excitation/Emission505/525
Label or DyepHrodo™ Green
Molecular Weight (g/mol)∼1000
Quantity3 x 100 μg
Reactive MoietyActive Ester, STP Ester
Shipping ConditionRoom Temperature
ColorGreen
Label TypepHrodo (pH sensitive)
Product LinepHrodo
Unit SizeEach

Frequently asked questions (FAQs)

Is there an alternative chaotrope for 8M guanidine-HCl solution used for determining degree of labeling (DOL) for pHrodo Dyes for Amine Labeling (Cat. Nos. P36010, P36011, P36012, P36013)?

Yes. A solution of 8 M urea in 20 mM HEPES can be used as an alternative for 8 M guanidine-HCl solution in the degree of labeling (DOL) determination protocol for pHrodo Dyes for Amine Labeling (Cat. Nos. P36010, P36011, P36012, P36013).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.