Owl™ P7-CST Multiple Gradient Gel Caster
Actual product may vary
Owl™ P7-CST Multiple Gradient Gel Caster
Cast one to five gels simultaneously with the Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
P7-SDSpacer, Thick
P7-10FGFrosted Blank Glass
P7-10FRFrosted Notched Glass
P7-10GBlank Glass
P7-10RNotched Glass
P7-10RANotched Alumina Plate
P7-CSTCaster
P7-SCSpacer, Thin
Catalog number P7-SD
HSN 39269099
Price (INR)
6,095.00
Each
Type:
Spacer, Thick
Price (INR)
6,095.00
Each
Cast one to five gels simultaneously with the Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster. One of the simplest devices to cast gradient gels, this caster is compatible with the Owl™ PBDS system.
  • With an external gradient maker, gels can be cast with ease and great reproducibility
  • The design and format of this unit will provide years of service in your lab
Specifications
DescriptionSpacer (2 sides,1 bottom)
For Use With (Equipment)Owl P7-CST System
Product LineOwl
TypeSpacer, Thick
Thickness (Metric)1.5 mm
Width (Metric)0.5 cm
Unit SizeEach