Elescan XRF-100 Online Multi-stream Elemental Slurry Analyzer
Elescan XRF-100 Online Multi-stream Elemental Slurry Analyzer
Elescan XRF-100 Online Multi-stream Elemental Slurry Analyzer
Elescan XRF-100 Online Multi-stream Elemental Slurry Analyzer
Thermo Scientific™
Elescan XRF-100 Online Multi-stream Elemental Slurry Analyzer
The Thermo Scientific Elescan XRF-100 Online Multi-stream Elemental Slurry Analyzer performs real-time analysis of slurry streams in mining and mineral processing.
Have Questions?
Catalog NumberType
PAXRFELESCAN100XRF Instrument
Catalog number PAXRFELESCAN100
Price (INR)
-
Type:
XRF Instrument
The Thermo Scientific Elescan XRF-100 Online Multi-stream Elemental Slurry Analyzer is a high-performance tool, designed for real-time analysis of slurry streams in mining and mineral processing. Using energy dispersive X-ray fluorescence (EDXRF), this advanced elemental slurry analyzer provides accurate measurement of metal concentrations of key process streams, aiding in in process control decision making and beneficiation circuit troubleshooting to minimize pay metal losses and optimize reagent usage.
A key advantage of combining EDXRF (single detector, many elements) and a low power air-cooled X-ray tube for continuous analysis is that a continuous X-ray energy spectrum.  This facilitates the detection of unexpected elements as the mine works through the ore deposit over time, enabling real-time slurry composition analysis with high precision. 
This combination allows the use of an economical low-power excitation source compared to WDXRF systems. With its long service life, replacement costs are a fraction of WDXRF excitation sources while providing excellent energy resolution and high counting rates with fast cycle times in a multi-stream elemental slurry analyzer.
Specifications
Sample TypeSample flow rates Recommended: 100–200 I/min Min–Max range: 70–250 I/min
Weight (Metric)Total system weight (12 lines) 1100 kg
TypeXRF Instrument
Unit SizeEach