Stratagene™ Mx3005P qPCR Instrument with Laptop and Software
Thermo Scientific™

Stratagene™ Mx3005P qPCR Instrument with Laptop and Software

Make results interpretation and reporting of mastitis-causing microorganisms easy with this qPCR instrument and software.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionVoltage
PF1449NStratagene Mx3005P qPCR Instrument with Laptop and Software110 V
PF1457NStratagene Mx3005P qPCR Instrument with Laptop and Software230 V
Catalog number PF1449N
Price (INR)
-
Description:
Stratagene Mx3005P qPCR Instrument with Laptop and Software
Voltage:
110 V

This product has been discontinued. The qPCR instrument of choice for Thermo Scientific™ PathoProof™ assays that target DNA in bovine milk samples is now the Applied Biosystems™ 7500 Fast qPCR instrument.

The instrument and laptop computer include a one-year warranty. The software changes the Ct-values to semi-quantitative results and reports the most abundant target if it is present in over 90% of the total bacterial load. The software license is for 1 to 5 users.

PathoProof RT-PCR assays identify the main mastitis-causing microorganisms and the Staphylococcal β-lactamase gene responsible for the penicillin resistance in Staphylococci. These microorganisms are responsible for 95 to 99% of all clinical and subclinical mastitis cases worldwide.

  • Assays are based on real-time PCR, an accurate and sensitive technology for identification of the target DNA in the sample
  • Tests can be used for analyzing fresh and preserved quarter milk samples, milk recording samples and bulk tank milk samples
  • All targeted species are detected simultaneously, overcoming the common problem of mixed growth results of bacterial culturing
  • Validation studies indicate superior analytical specificity and sensitivity over traditional methods, even for samples that show no growth in culture
  • Accurate detection of viable, dead and growth-inhibited microorganisms reduces no-growth results

Not all products are available for sale in all territories. Please inquire.

Permissible use. This product is intended to be used for the purpose of detection and/or analysis of microorganisms in milk for quality assurance and quality control purposes (Food Testing Applications), as well as for identification, enumeration, or count of microorganisms in raw material sample, process control sample, or finished product sample of an industrial process for the purpose of detecting the presence, absence or amount either of a contaminant or of an intended component (Industrial Microbiology Applications). Not for use in veterinary diagnostic procedures. The quality system of Thermo Scientific PathoProof is certified according to standard SFS-EN ISO9001:2008.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
DescriptionStratagene Mx3005P qPCR Instrument with Laptop and Software
DetectionMastitis-causing Microorganisms
Excitation350 to 750 nm
For Use With (Application)Mastitis testing in Bovine Milk Samples
For Use With (Equipment)Real Time PCR System
Height (English)18 in.
Thermal Range25°C to 99°C
Thermal Uniformity+/- .25°C at 72°C
Voltage110 V
Weight20.4 kg (45 lb.)
Width (English)14 in.
Block FormatSolid-state, Peltier-based
Unit SizeEach
Contents & Storage
Laptop, Software, 1 Year Warranty