Product Image
Invitrogen™

Cytochalasin D

Cytochalasin D is a cell permeable fungal toxin that binds to the barbed end of actin filaments inhibiting both theRead more
Have Questions?
Catalog NumberQuantity
PHZ10631 mg
Catalog number PHZ1063
Price (INR)
14,498.00
온라인 행사
Ends: 30-Sep-2026
26,360.00
Save 11,862.00 (45%)
Each
Quantity:
1 mg
Price (INR)
14,498.00
온라인 행사
Ends: 30-Sep-2026
26,360.00
Save 11,862.00 (45%)
Each
Cytochalasin D is a cell permeable fungal toxin that binds to the barbed end of actin filaments inhibiting both the association and dissociation of subunits. This compound causes the disruption of actin filaments and inhibition of actin polymerization.

Format/Formulation: Lyophilized
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gene ID (Entrez)60
Inhibitor PropertiesChemical
InhibitorsCytoskeletal Inhibitor
Molecular Weight (g/mol)507.6
Product TypeCytochalasin D
Quantity1 mg
Shipping ConditionDry Ice
Specific Inhibit or TargetActin
SterilityNon-sterile
FormSolid
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C).