Product Image
Invitrogen™

LY294002

LY294002
Have Questions?
Catalog NumberQuantity
PHZ11445 mg
Catalog number PHZ1144
Price (INR)
-
Quantity:
5 mg

LY294002 is a potent and specific cell-permeable inhibitor of phosphatidylinositol 3-kinase (PI3-kinase).

Format/Formulation: Lyophilized

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Gene ID (Entrez)5294
Inhibitor PropertiesChemical
InhibitorsKinase Inhibitor (PI 3-Kinase Inhibitor), Kinase Inhibitor
Molecular Weight (g/mol)307.4
Product TypeInhibitor
Purification MethodTLC
Quantity5 mg
Shipping ConditionDry Ice
Specific Inhibit or TargetPI3-Kinase
SterilityNon-sterile
FormSolid
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).