Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine
Actual product may vary
Thermo Scientific™

Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine

Carry out environmental monitoring in your most regulated environments using Thermo Scientific™ Triple Wrapped Plates for passive and active air sampling applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatMedia TypeManufacturing Origin
PO5511D100/Pk.55mm Contact PlateTryptone Soya AgarGermany
PO5501B100/Pk.90mm MonoplateTryptone Soya AgarGermany
Catalog number PO5511D
Price (INR)
-
Quantity:
100/Pk.
Format:
55mm Contact Plate
Media Type:
Tryptone Soya Agar
Manufacturing Origin:
Germany
Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar plates, complete with VHP indicator, Lecithin, Polysorbate 80, Sodium Thiosulphate and L-Histidine.
Achieve confidence in your results and ensure optimum media fertility using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.

Three protective layers reduce risk of contamination:
  • Primary film with moisture control patch
  • Secondary film VHP barrier with immobilized desiccant sachet
  • Tertiary film dust cover

The incorporation of novel elements enhance moisture control by permitting the passage and absorption of excess moisture
 
Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs

Not all products are available for sale in all territories. Please inquire.
Specifications
ColorIvory, transparent
Fill Weight13.0g ± 0.5g
Format55mm Contact Plate
Media TypeTryptone Soya Agar
Packaging TypeTriple Wrapped - Irradiated
Quantity100/Pk.
Shelf Life43 Weeks
Manufacturing OriginGermany
Product TypeAgar
Unit SizeEach
Contents & Storage
2 to 25°C