PoroMetric
Thermo Scientific™
PoroMetric
The Phenom Desktop SEM with PoroMetric software allows easy generation and analysis of SEM images. The integrated PoroMetric software allows the user to gather data on distribution of pores, and pore parameters like pore size and aspect ratio.
Have Questions?
Catalog number POROMETRIC
Price (INR)
-
The Thermo Scientifc™ Phenom™ Desktop SEM with Thermo Scientific™ PoroMetric™ software allows easy generation and analysis of SEM images. The integrated PoroMetric software allows the user to gather data on distribution of pores, and pore parameters like pore size and aspect ratio.
Key Benefits​
  • Acquire images directly from the Phenom
  • Correlate pore features such as area, aspect ratio, major and minor axis
  • Fast and convenient operation improves workflow and makes scheduling simple and predictable
  • Image collection is limitless as digital files are easily stored on a network or USB disk for data sharing, communication, or later reference
  • Statistical data with high-quality images