Powder Sampling Cups and Vials for Antaris™ II for Antaris II NIR
Thermo Scientific™
Powder Sampling Cups and Vials for Antaris™ II for Antaris II NIR
Thermo Scientific Sampling Cups and Vials provide consistent sample presentation to ensure reproducible diffuse reflectance measurements.
Have Questions?
Catalog number POWDERCUPS
Price (INR)
-
Thermo Scientific Sampling Cups and Vials provide consistent sample presentation to ensure reproducible diffuse reflectance measurements. Sampling Cups and Vials allow easy, reproducible diffuse reflectance measurements for powders, grains and granular solid samples using the Integrating Sphere module of the Antaris II™ FT-NIR analyzer.
Sampling Cup Features
  • Low OH quartz window
  • Internal plunger mechanism maintains consistent powder packing
  • Multiple sampling areas that accommodate a wide variety of sample types—sample areas of 1, 3, 5, and 12cm
  • Optional Positioning Ring Kit includes two centering rings, one off-center ring
Sample Vial Features
  • Glass vials available in two sizes
  • 19mm vials have 7.4mL capacity (approximately 2 dram)
  • 12mm vials have 1.8mL capacity (approximately 0.5 dram)
  • Optional Vial Holder Kit includes one disc to center 19mm vial, one disc to center 12mm vial over the Integrating Sphere
Recommended for:
  • Powders
  • Solids
Specifications
DescriptionPowder Sampling Cups and Vials for Antaris II NIR
Sample TypePowders, Solids, Pellets, Grains, Feed, Food products
For Use With (Equipment)Antaris II Integrating Sphere
TypeSampling Cups
Unit SizeEach