KingFisher™ PlasmidPro Maxi Processor
KingFisher™ PlasmidPro Maxi Processor
KingFisher™ PlasmidPro Maxi Processor
KingFisher™ PlasmidPro Maxi Processor
Thermo Scientific™

KingFisher™ PlasmidPro Maxi Processor

The KingFisher™ PlasmidPro Maxi Processor transforms the maxi-scale plasmid purification process with a cutting-edge, fully automated workflow that eliminates labor-intensive centrifugation and pipetting steps.
Have Questions?
Catalog NumberQuantity
PPMX10001 Ea.
Catalog number PPMX1000
Price (INR)
-
Quantity:
1 Ea.

The KingFisher PlasmidPro Maxi Processor transforms the maxi-scale plasmid purification process with a cutting-edge, fully automated workflow that eliminates labor-intensive centrifugation and pipetting steps. Simply pour in the culture and let the instrument handle the rest. In under 75 minutes, this innovative system efficiently executes all essential purification steps, delivering high-quality, endotoxin-free plasmid DNA.

True walkaway automation

The KingFisher PlasmidPro system is fully automated, eliminating the need for pipetting or centrifugation steps. It completes each run in under 75 minutes, allowing you to focus on other tasks.

Exceptional quality

Optimized for maxi-scale plasmid DNA purification from 150 mL of bacterial culture, the system consistently delivers high-quality, advanced, transfection-grade plasmid DNA.

Convenient pre-filled cartridge

The one-piece, pre-filled KingFisher PlasmidPro Maxi Processor Endotoxin-Free Cartridge eliminates setup time and ensures a simple clean-up process, enabling quick turnaround for your next purification.

Effortless installation and setup

The installation process can be completed in minutes on any benchtop, ensuring immediate usability.

Optional AB Assurance Plan

The AB Assurance Plan is a comprehensive repair plan that aims to optimize instrument performance and ensure the availability of critical systems. It includes preventive maintenance, proactive instrument monitoring, and repairs, all aimed at keeping your laboratory operating smoothly.

Additional details

The KingFisher PlasmidPro Maxi Processor is specifically optimized for cultures grown in Luria-Bertani (LB) medium.

At the end of the procedure, the system delivers endotoxin-free plasmid DNA resuspended in TE buffer, ensuring optimal quality and compatibility with downstream applications.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dimensions19.7 x 11.8 x 23.6 in. (50 x 30 x 60 cm)
Display7 in. Touchscreen
Electrical Requirements100/240 VAC, 50/60 Hz (nominal), automatic voltage detection
For Use With (Application)Plasmid purification, advanced transfection applications
Frequency50/60 Hz
Memory30 GB
PortsUSB
Purification TargetPlasmid DNA
Quantity1 Ea.
Run Time<75 min.
Sample Volume150 mL
Samples per Run1 Sample/Run
Shipping ConditionRoom temperature
Capacity1 Sample/Run
FormatCartridge
Plates per DeckNone
Product TypePlasmid Prep
Sample TypeBacteria
Suitable Downstream AnalysisAdvanced Transfection Applications
Volume (Metric) Processing150 mL
Unit SizeEach