ProGC+ Laboratory and Process Gas Chromatograph
Thermo Scientific™
ProGC+ Laboratory and Process Gas Chromatograph
The Thermo Scientific™ ProGC+ Laboratory and Process Gas Chromatograph has been discontinued; however, we continue to provide services and support for this product.
Have Questions?
Catalog NumberProduct Type
PROGCANLYZGas Chromatograph
Catalog number PROGCANLYZ
Price (INR)
-
Product Type:
Gas Chromatograph
The Thermo Scientific™ ProGC+ Laboratory and Process Gas Chromatograph has been discontinued; however, we continue to provide services and support for this product.
The comprehensive range of optimized high-sensitivity detectors includes thermal conductivity, flame ionization, electron capture, and the ultra-trace pulsed discharge detector. All of the ionization detectors are fully interchangeable, sharing a common base body. Instrument control is exceptionally easy, using the industry-standard user interface, with additional dedicated keys to speed method setup. The flexible, 90-position autosampler facilitates the programming of a wide range of automatic injection techniques, providing optimum simultaneous or sequential dual injection option. The column oven provides superior performance in oven heating, cooling and thermal stability. The range of leading-edge injectors include split/splitless, cold on-column, PTV, and packed injectors.
Features:
  • Optimized column oven with fast heating/cooling rates
  • Comprehensive electronic gas controls
  • Leading-edge injector designs
  • Interchangeable ionization detectors
  • Industry-standard user-interface
  • High performance autosampler
  • Versatile valve automation
  • Powerful, flexible GC software
  • Outstanding serviceability
  • Widest range of application-guaranteed analyzer systems
  • Full digital pressure or flow control is provided for all carrier and detector gases
Recommended for:
  • Laboratory quality control, research and development
  • Industrial and electronic gases (UHP H2, He, O2, N2, Ar, CO2, HCl, HBr, Cl2, BCl3, NF3, WF6 etc.)
  • Natural gas by ASTM, GPA, ISO standard methods (e.g., N2, O2, CO2, C1 to C6 or C1 to C12; sulfur odorants)
  • Petroleum refining (refinery gas, LPG, capillary PIANO, oxygenates, SimDis)
  • Petrochemicals (trace CO/CO2, Cl2, formaldehyde, ethylene, propylene etc.)
  • Power generation (transformer oil, nuclear reactor gas)
  • More applications in steel, chemicals, environmental, etc.
Specifications
DescriptionProGC+ Laboratory and process gas chromatograph
Product TypeGas Chromatograph
Unit SizeEach