Product Image
Gibco™

Bovine Fetuin (custom product)

This is a GIBCO Custom Product. Please complete and submit the Custom Media Inquiry Form and a Customer Service RepresentativeRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PSB10052IJ10 g, 10 g, 10 g, 10 g
PSB10052II1 g, 1 g, 1 g, 1 g
PSB10054II1 g, 1 g, 1 g, 1 g
PSB10052IC5 g, 5 g, 5 g, 5 g
PSB10054IC5 g, 5 g, 5 g, 5 g
1034403410 g, 10 g, 10 g, 10 g
PSB10054IJ10 g, 10 g, 10 g, 10 g
10344042100 g
PSB10052IM100 g, 100 g, 100 g, 100 g
PSB10054IM100 g
Catalog number PSB10052IJ
Price (INR)
-
Quantity:
10 g, 10 g, 10 g, 10 g
This is a GIBCO Custom Product. Please complete and submit the Custom Media Inquiry Form and a Customer Service Representative will contact you regarding pricing and availability.
Specifications
Gene ID (Entrez)504615
Quantity10 g, 10 g, 10 g, 10 g
Shipping ConditionRoom Temperature, Room Temperature, Room Temperature, Room Temperature
SpeciesBovine
FormSolid, Solid, Solid, Solid
SpeciesBovine
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).