Fluorescein-IKK Substrate

Fluorescein-IKK Substrate

Fluorescein-IKK Substrate
Have Questions?
Catalog NumberQuantity
PV35071 mg
Catalog number PV3507
Price (INR)
132,523.00
Each
Quantity:
1 mg
Price (INR)
132,523.00
Each
A fluorescein-labeled peptide substrate that is primarily used with the IkappaB kinase (IKK) family.

For use in combination with the LanthaScreen™ Tb-IkappaBalpha [pSer32] antibody (PV3562) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Amino Acid SequenceFluorescein- DRHDSGLDSMKDE
Assay EntryBiochemical terbium phospho-specific activity
Gene ID (Entrez)1147, 2869, 3551
Label or DyeFITC (Fluorescein)
Molecular Weight (g/mol)1864 Da
Quantity1 mg
Shipping ConditionDry Ice
SubstrateFluorescein-IKK
Target EntryIKBKE, CHUK, GRK5, GRK6, TBK1, IKBKB
Validated KinasesCHUK, GRK5, GRK6, IKBKB, IKBKE
Detection MethodFluorescence
Substrate PropertiesPeptide-Based Substrate
Substrate TypeKinase Substrate
Target EnzymeSerine/Threonine Kinases
Unit SizeEach
Contents & Storage
1 mg/mL fluorescein-IKK substrate in water. Store at -20°C and protect from light.