Product Image

Fluorescein-Poly GT

A general fluorescein peptide substrate consisting of a polymer of glutamate, and tyrosine that is used to monitor tyrosine kinaseRead more
Have Questions?
Catalog NumberQuantity
PV36101 mL
Catalog number PV3610
Price (INR)
63,804.00
Offre exceptionnelle en ligne
Ends: 31-Mar-2026
106,340.00
Save 42,536.00 (40%)
Each
Quantity:
1 mL
Price (INR)
63,804.00
Offre exceptionnelle en ligne
Ends: 31-Mar-2026
106,340.00
Save 42,536.00 (40%)
Each
A general fluorescein peptide substrate consisting of a polymer of glutamate, and tyrosine that is used to monitor tyrosine kinase activity.

For use in combination with the LanthaScreen™ Tb-pY20 Antibody (PV3552) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Amino Acid SequenceFluorescein- Poly Glu:Tyr (4:1)
Assay EntryBiochemical terbium phospho-specific activity
Label or DyeFITC (Fluorescein)
Quantity1 mL
Shipping ConditionDry Ice
SubstrateFluorescein-Poly GT
Target EntryABL1 G250E, CSF1R, CSK, EPHA5, KIT, PDGFRA T674I, PDGFRA V561D, PTK6, RET Y791F, SYK, TYRO3, ZAP70, FGFR1, FGFR2, BLK, EGFR, EPHB2, FGFR4, IGF1R, LYNA, MATK, MERTK, NTRK3, RET V804L, ROS1, TEK, TTK, FLT1, MUSK, SRCN1, TEC, ABL1 E255K, BMX, EPHA4, FYN, PTK2, PTK2B, RET, ABL1 T315I, EPHA3, EPHB4, FLT3, FLT4, INSR, LCK, YES1, EGFR L858R, ERBB4, KDR, MET, TNK2, ABL1, ABL2, AXL, FGFR3, FGFR3 K650E, FGR, HCK, KIT V654A, EPHA1, EPHA8, EPHB3, INSRR, LYNB, MST1R, NTRK1, PDGFRA D842V, SRMS, EGFR L861Q, EPHA2, FER, LTK, PDGFRA, PDGFRB, EGFR T790M, EPHA7, EPHB1, MET M1250T, FRK, ITK, FLT3 D835Y, SRC, ABL1 Y253F, BTK, DDR2, FES, KIT T670I, TXK, ERBB2, NTRK2
Validated KinasesABL1, ABL2, AXL, BLK, BMX, BTK, CSF1R, CSK, DDR2, EGFR, EPHA1, EPHA2, EPHA3, EPHA4, EPHA5, EPHA7, EPHA8, EPHB1, EPHB2, EPHB3, EPHB4, ERBB2, ERBB4, FER, FGFR1, FGFR2, FGFR3, FGFR4, FGR, FLT1, FLT3, FLT4, FRK, FYN, HCK, IGF1R, INSR, KDR, KIT, LCK, LTK, LYNA, LYNB, MATK, MERTK, MET, MST1R, MUSK, NTRK1, NTRK2, NTRK3, PDGFRA, PDGFRB, PTK2, PTK2B, PTK6, RET, ROS1, SRC, SRCN1, SRMS, SYK, TEC, TEK, TNK2, TTK, TYRO3, YES1, ZAP70
Detection MethodFluorescence
FormLiquid
Substrate PropertiesPeptide-Based Substrate
Substrate TypeKinase Substrate
Target EnzymeTyrosine Kinases
Unit SizeEach
Contents & Storage
30 μM Fluorescein Poly-GT peptide substrates in 10 mM HEPES, 137 mM NaCl, 2.7 mM KCl (HEPES buffered saline). Store at -20°C and protect from light.