Biotin-Ubiquitin
Invitrogen™

Biotin-Ubiquitin

Biotin-labeled ubiquitin for use as a general research tool, or in high throughput assays to monitor protein ubiquitination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PV4380100 μg
PV437910 μg
Catalog number PV4380
Price (INR)
141,966.00
キャンペーン価格
Ends: 30-Sep-2026
189,286.70
Save 47,320.70 (25%)
Each
Quantity:
100 μg
Price (INR)
141,966.00
キャンペーン価格
Ends: 30-Sep-2026
189,286.70
Save 47,320.70 (25%)
Each

Biotin-labeled ubiquitin for use as a general research tool, or in high throughput assays to monitor protein ubiquitination. Due to the method in labeling the ubiquitin protein, all of the lysines with the protein are available for further conjugation to form poly-ubiquitin chains.

For use in combination with other LanthaScreen™ products including LanthaScreen™ Tb-Ubiquitin (Cat. No. PV4375), Fluorescein-Ubiquitin (PV4377), or a LanthaScreen™ antibody such as Tb-anti-biotin (PV4749).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Excitation/Emission340/490, 520 nm
For Use With (Equipment)Microplate Reader
Label TypeBiotin & Other Haptens
Label or DyeBiotin
Purity or Quality Grade≥90% by SDS-PAGE
Quantity100 μg
ReadoutEnd Point
Shipping ConditionDry Ice
Substrate PropertiesProtein-Based Substrate
TechniqueTR-FRET
ConjugateBiotin
FormLiquid
Unit SizeEach
Contents & Storage
Biotin labeled ubiquitin in HEPES buffered saline. Store at -20°C.

Citations & References (1)

Citations & References
Abstract
TR-FRET-based high-throughput screening assay for identification of UBC13 inhibitors.
Authors:Madiraju C, Welsh K, Cuddy MP, Godoi PH, Pass I, Ngo T, Vasile S, Sergienko EA, Diaz P, Matsuzawa S, Reed JC,
Journal:J Biomol Screen
PubMed ID:22034497
UBC13 is a noncanonical ubiquitin conjugating enzyme (E2) that has been implicated in a variety of cellular signaling processes due to its ability to catalyze formation of lysine 63-linked polyubiquitin chains on various substrates. In particular, UBC13 is required for signaling by a variety of receptors important in immune regulation, ... More