Phosphate Sensor

Phosphate Sensor

The Phosphate Sensor is a highly sensitive reagent for the detection of inorganic phosphate, either in real-time kinetic or end-pointRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PV440610 nmol
PV4407100 nmol; 10,000 assay(s)
Catalog number PV4406
Price (INR)
-
Quantity:
10 nmol
The Phosphate Sensor is a highly sensitive reagent for the detection of inorganic phosphate, either in real-time kinetic or end-point mode. This product is a purified form of recombinant E. coli phosphate-binding protein labeled with the fluorophore MDCC, which is sensitive to changes in its environment. Binding of inorganic phosphate by Phosphate Sensor is rapid and tight (Kd ∼ 0.1 μM), resulting in a large increase in fluorescence. The Phosphate Sensor detects phosphate in the high-nanomolar to low-micromolar range, which is 100-fold more sensitive than malachite green. This reagent provides you with a sensitive assay for detecting enzymes that produce inorganic phosphate either directly or indirectly through coupled reactions, such as phosphatases, ATPases, GTPases, and phosphodiesterases.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayPhosphatase Assay
Detection MethodFluorescence
Excitation/Emission430⁄450
For Use With (Application)Fluorescence Intensity
For Use With (Equipment)Microplate Reader
Format384-well plate
Label or DyeMDCC
Product TypePhosphate Sensor
Quantity10 nmol
ReadoutKinetic
Shipping ConditionDry Ice
TargetGTPases, ATPases, Phosphatases, Phosphodiesterases
Unit SizeEach
Contents & Storage
Phosphate Sensor is supplied as a 500 μM solution in 10 mM Tris (pH 7.6) and 50 mM NaCl. Store product at -80°C.