GFP-ATF2 (19-96)

GFP-ATF2 (19-96)

GFP-ATF2 (19-96)
Have Questions?
Catalog NumberQuantity
PV444520 nmol
Catalog number PV4445
Price (INR)
102,785.00
Online offer
Ends: 30-Sep-2026
137,046.30
Save 34,261.30 (25%)
Each
Quantity:
20 nmol
Price (INR)
102,785.00
Online offer
Ends: 30-Sep-2026
137,046.30
Save 34,261.30 (25%)
Each
A truncated version (residues 19-96) of ATF2 has been expressed as a fusion with GFP (Green Fluorescent Protein) to act as a substrate for the JNK and p38 families of kinases.

For use in combination with the LanthaScreen™ Tb-anti-ATF2 [pThr71] antibody (PV4451) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay EntryBiochemical terbium phospho-specific activity
Label or DyeGFP (EmGFP)
Molecular Weight (g/mol)478000 Da
Quantity20 nmol
Shipping ConditionDry Ice
SubstrateATF2 (19-96)
Target EntryMAPK13, MAPK8, MAPK9, MAPK14, MAPK3, MAPK1, MAPK11, MAPK12, MAPK10
Validated KinasesMAPK1, MAPK3, MAPK8, MAPK9, MAPK10, MAPK11, MAPK12, MAPK13, MAPK14
Detection MethodFluorescence
FormLiquid
Substrate PropertiesProtein-Based Substrate
Substrate TypeKinase Substrate
Target EnzymeSerine/Threonine Kinases
Unit SizeEach
Contents & Storage
GFP-ATF2 protein substrate in 10 mM Tris, pH 7.5, 150 mM NaCl, 10% glycerol, 1mM DTT. Store at -80°C.