Product Image
Thermo Scientific™

GFP-4E-BP1

4E-BP1 expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for FRAP1 (mTor).
Have Questions?
Catalog NumberQuantity
PV475920 nmol
Catalog number PV4759
Price (INR)
100,208.00
Each
Quantity:
20 nmol
Price (INR)
100,208.00
Each

4E-BP1 expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for FRAP1 (mTor).

For use in combination with the LanthaScreen™ Tb-anti-4E-BP1 [pThr46] Antibody (Cat. No. PV4757) to evaluate inhibitors in medium to high throughput screening applications. An optimized mTor assay buffer (PV4794) is also available.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay EntryBiochemical terbium phospho-specific activity
Label or DyeGFP (EmGFP)
Molecular Weight (g/mol)45,600 Da
Quantity20 nmol
Shipping ConditionDry Ice
Substrate4E-BP1
Target EntrymTOR, FRAP1
Validated KinasesFRAP1 (mTOR), FRAP1 (mTOR)
Detection MethodFluorescence
FormLiquid
Substrate PropertiesProtein-Based Substrate
Substrate TypeKinase Substrate
Target EnzymeSerine/Threonine Kinases
Unit SizeEach
Contents & Storage
GFP-4E-BP1 protein substrate in 10 mM Tris, pH 7.5, 150 mM NaCl, 10% glycerol, 2 mM DTT

Store at -80°C.