Product Image

Fluorescein-MAP2K1

Inactive MAP2K1 (MEK1) has been labeled with fluorescein, and is a substrate for MAPK family members.For use in combination withRead more
Have Questions?
Catalog NumberQuantity
PV481220 nmol
Catalog number PV4812
Price (INR)
113,057.00
Online offer
Ends: 31-Mar-2026
188,428.00
Save 75,371.00 (40%)
Each
Quantity:
20 nmol
Price (INR)
113,057.00
Online offer
Ends: 31-Mar-2026
188,428.00
Save 75,371.00 (40%)
Each
Inactive MAP2K1 (MEK1) has been labeled with fluorescein, and is a substrate for MAPK family members.

For use in combination with the LanthaSreen™ Tb-MAP2K1 [pSer217/221] Antibody (PV4813) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay EntryBiochemical terbium phospho-specific activity
Gene ID (Entrez)10454, 10746, 1326, 4215, 4217, 4293, 4294, 4296, 5894, 673, 6885
Label or DyeFITC (Fluorescein)
Molecular Weight (g/mol)50000 Da
Quantity20 nmol
Shipping ConditionDry Ice
SubstrateMAP2K1
Target EntryMAP3K2, RAF1 (cRAF) Y340D Y341D, MAP3K10, MAP3K9, MAP3K7/MAP3K7IP1, MAP3K5, MAP3K11, BRAF V599E, BRAF, MAP3K8, MAP3K3
Validated KinasesBRAF, MAP3K2, MAP3K3, MAP3K5, MAP3K7, MAP3K8, MAP3K9, MAP3K10, MAP3K11
Detection MethodFluorescence
Substrate PropertiesProtein-Based Substrate
Substrate TypeKinase Substrate
Target EnzymeSerine/Threonine Kinases
Unit SizeEach
Contents & Storage
Fluorescein-MAP2K1 protein substrate in 10 mM Tris, pH 7.5, 150 mM NaCl, 10% glycerol, 1mM DTT. Store at -80°C.