ZIPtide Substrate
Thermo Scientific™

ZIPtide Substrate

A peptide substrate for the Ca/Calmodulin-dependent protein kinase (CaM) family including CAMK1 and DAPK1. For use with the Adapta™ UniversalRead more
Have Questions?
Catalog NumberQuantity
PV50941 mL
Catalog number PV5094
Price (INR)
139,082.00
Each
Quantity:
1 mL
Price (INR)
139,082.00
Each
A peptide substrate for the Ca/Calmodulin-dependent protein kinase (CaM) family including CAMK1 and DAPK1. For use with the Adapta™ Universal Kinase Assay (PV5099).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Amino Acid SequenceKKLNRTLSFAEPGW
Assay EntryBiochemical ADP accumulation
Gene ID (Entrez)1612, 8536
Molecular Weight (g/mol)1646.9 Da
Quantity1 mL
Shipping ConditionDry Ice
Substratepeptide for CaM kinases
Target EntryDAPK1, CAMK1
Validated KinasesCAMK1, DAPK1
Detection MethodFluorescence
Substrate PropertiesPeptide-Based Substrate
Substrate TypeKinase Substrate
Target EnzymeSerine/Threonine Kinases
Unit SizeEach
Contents & Storage
2 mM ZIPtide Peptide substrate in 50 mM HEPES (pH 7.5), 0.01% Brij-35, 10 mM MgCl2 and 1 mM EGTA. Store (at -20°C.)