Product Image
Invitrogen™

Alexa Fluor™ 647-Poly-GT Peptide Substrate

A general AlexaFluor647-labeled peptide substrate consisting of a polymer of glutamate, and tyrosine that is used to monitor tyrosine kinase acitivty.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PV56931 nmol
PV583610 nmol
Catalog number PV5693
Price (INR)
26,618.00
Online offer
Ends: 30-Sep-2026
48,396.00
Save 21,778.00 (45%)
Each
Quantity:
1 nmol
Price (INR)
26,618.00
Online offer
Ends: 30-Sep-2026
48,396.00
Save 21,778.00 (45%)
Each

A general AlexaFluor647-labeled peptide substrate consisting of a polymer of glutamate, and tyrosine that is used to monitor tyrosine kinase acitivty.

For use in combination with the LanthaScreen™ Eu-PY20 Antibody (Cat. No. PV5696) to evaluate inhibitors in medium to high throughput screening applications.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay EntryBiochemical europium phospho-specific activity
ColorFar-Red
Excitation/Emission340/665 nm
Label or DyeAlexa Fluor™ 647
Product LineLanthaScreen
Quantity1 nmol
Shipping ConditionDry Ice
SubstrateRandom (Glu:Tyr) polymer in a ratio of 4:1
Target EntryABL1 G250E, CSF1R, CSK, EPHA5, KIT, PDGFRA V561D, SYK, TYRO3, ZAP70, FGFR1, FGFR2, EGFR, EPHB2, FGFR4, IGF1R, LYNA, MATK, MERTK, NTRK3, RET V804L, TEK, FLT1, MUSK, SRCN1, ABL1 E255K, EPHA4, FYN, PTK2, PTK2B, RET, ABL1 T315I, EPHA3, EPHB4, FLT3, FLT4, INSR, LCK, YES1, EGFR L858R, ERBB4, KDR, MET, TNK2, ABL1, ABL2, AXL, FGFR3, FGR, HCK, KIT V654A, EPHA1, EPHA8, EPHB3, INSRR, LYNB, NTRK1, PDGFRA D842V, SRMS, EGFR L861Q, EPHA2, FER, LTK, PDGFRA, EPHA7, EPHB1, FRK, ITK, FLT3 D835Y, SRC, BTK, FES, ERBB2, NTRK2
TechniqueTR-FRET
Validated KinasesEGFR
Detection MethodFluorescence
FormLiquid
Substrate PropertiesPeptide-Based Substrate
Substrate TypeKinase Substrate
Target EnzymeTyrosine Kinases
Unit SizeEach
Contents & Storage
30 μM AlexaFluor 647 Poly-GT peptide substrate in 10 mM HEPES, 137 mM NaCl, 2.7 mM KCl, pH 7.5 (HEPES buffered saline).

Store at -20°C and protect from light.