Product Image
Nuclear Receptor Buffer K
Nuclear Receptor Buffer K
Have Questions?
Catalog NumberQuantity
PV604325 mL
Catalog number PV6043
Price (INR)
24,260.00
Each
Quantity:
25 mL
Price (INR)
24,260.00
Each
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Recommended StorageTR-FRET Coregulator Buffer K may be shipped frozen or at room temperature. It should be stored at room temperature (20-30°C).
GradeResearch Grade
Physical FormLiquid
Product LineLanthaScreen
Quantity25 mL
Unit SizeEach