5X Kinase Buffer A
Invitrogen™

5X Kinase Buffer A

The 5X Kinase Buffer A is a concentrated general kinase assay buffer that can be used with a variety ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PV6135100 ml
PV31894 mL
Catalog number PV6135
Price (INR)
88,004.00
Each
Quantity:
100 ml
Price (INR)
88,004.00
Each
The 5X Kinase Buffer A is a concentrated general kinase assay buffer that can be used with a variety of assay technologies such as, LanthaScreen™, Adapta™, and Z’-LYTE™ assays.

Ready to use and validated with LanthaScreen™, Adapta™, and Z’LYTE™ assays

• Optimized 5X concentrated formulation
• Simplified handling with room temperature storage

Easily Screen for Kinase Targets using an Invitrogen™ Platform
For more information on the kinase assays that can be used with our 5X Kinase Buffer A see the following links:

LanthaScreen™ Eu Binding Assay
LanthaScreen™ Activity Assay
Adapta™ Universal Kinase Assay
Z´-LYTE™ Kinase Assay Kits

Save Freezer Space
The 5X Kinase Buffer can be stored at room temperature, saving you time and precious freezer space.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialKinase Buffer A
Includes250 mM HEPES (pH 7.5), 50 mM MgCl2, 5 mM EGTA, and 0.05% Brij-35
Concentration5X
For Use With (Application)Pharma & Biopharma; Protein Assays and Analysis
For Use With (Equipment)Adapta™ and Z-LYTE™ assays;
Physical FormLiquid
Quantity100 ml
pH7.5
Unit SizeEach