Ethyl acetate, GMP, Multi-Compendial
Greenfield Global

Ethyl acetate, GMP, Multi-Compendial

This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
Have Questions?
Catalog number PZP3300000EPDM55M
Price (INR)
-
This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes. It is not intended for use as an active ingredient in drug manufacturing, nor as a medical device or disinfectant. Appropriate/legal use of this product is the responsibility of the user.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point77°C
CAS141-78-6
Chemical Name or MaterialEthyl Acetate
ColorColorless
Molecular FormulaC4H8O2
PackagingMetal drum
SynonymAcetic acid ethyl ester
GradeMulti-Compendial
Molecular Weight (g/mol)88.11
Percent Purity100%
Quantity55 gal.
Unit Size55 gal.