Aluminum Ammonium Sulphate (ER ACS), Qualigens
Qualigens

Aluminum Ammonium Sulphate (ER ACS), Qualigens

Have Questions?
Catalog NumberQuantityPackaging
Q11085500 gPlastic Bottles
Catalog number Q11085
HSN 28333010
Price (INR)
811.00
Each
Quantity:
500 g
Packaging:
Plastic Bottles
Request bulk or custom format
Price (INR)
811.00
Each
Specifications
Boiling Point200°C
Melting Point93.5°C
CAS7784-26-1
ChemAlert Storage SymbolGray
pH3 Min (5% w/v solution in carbon dioxide free water)
View more

Detailed specifications PDF attached below