Oxalic Acid (ER ACS), Qualigens
Qualigens

Oxalic Acid (ER ACS), Qualigens

Have Questions?
Catalog NumberQuantityPackaging
Q13195500 gPlastic Bottles
Catalog number Q13195
HSN 29171110
Price (INR)
886.00
Each
Quantity:
500 g
Packaging:
Plastic Bottles
Request bulk or custom format
Price (INR)
886.00
Each
Specifications
CAS6153-56-6
ChemAlert Storage SymbolWhite
Melting Point98°C
Health Hazard 2DANGER!
Health Hazard 3Emergency Overview
Causes severe eye burns. Harmful in contact with skin and if swallowed. Irritating to respiratory system. Corrosive to metals. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:1
Instability:0
View more

Detailed specifications PDF attached below