You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Biotinylated Positive Control Oligo Probe (ALU)
Thermo Scientific™
Biotinylated Positive Control Oligo Probe (ALU)
Biotinylated Positive Control Oligo Probe
(ALU)
Read more
Have Questions?
Catalog Number
Quantity
Q151P.0100
1 mL
Catalog number
Q151P.0100
Price (INR)
51,752.00
Each
Add to cart
Quantity:
1 mL
Price (INR)
51,752.00
Each
Add to cart
Biotinylated Positive Control Oligo Probe (ALU)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or Dye
Biotin
Product Type
Oligonucleotides
Quantity
1 mL
Shipping Condition
Wet Ice
Product Line
REMBRANDT
Unit Size
Each
Contents & Storage
Store in refrigerator (2°C to 8°C).