Magnesium Chloride (ER ACS), Qualigens
Qualigens

Magnesium Chloride (ER ACS), Qualigens

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
50 kg
Packaging:
View all
Plastic Bottles
Catalog number Q15535
HSN 28273100
Price (INR)
635.00
Each
Quantity:
500 g
Packaging:
Plastic Bottles
Request bulk or custom format
Specifications
Melting Point117°C
CAS7791-18-6
ChemAlert Storage SymbolGray
Health Hazard 2WARNING!
Health Hazard 3Emergency Overview
Irritating to eyes and respiratory system. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:1
View more

Detailed specifications PDF attached below