Zinc Dust (ER), Qualigens
Qualigens

Zinc Dust (ER), Qualigens

Have Questions?
Catalog NumberQuantityPackaging
Q15995500 gPlastic Bottles
Catalog number Q15995
HSN 79031000
Price (INR)
2,137.00
Each
Quantity:
500 g
Packaging:
Plastic Bottles
Request bulk or custom format
Price (INR)
2,137.00
Each
Specifications
CAS7440-66-6
ChemAlert Storage SymbolGray
Melting Point419°C
Boiling Point907°C
Health Hazard 2CAUTION!
View more

Detailed specifications PDF attached below