Cyclohexanone (ER ACS), Qualigens
Cyclohexanone (ER ACS), Qualigens
Qualigens

Cyclohexanone (ER ACS), Qualigens

Have Questions?
Catalog NumberQuantityPackaging
Q18235500 mLGlass Bottle
Catalog number Q18235
HSN 29142200
Price (INR)
2,740.00
Each
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
2,740.00
Each
Add to cart
Specifications
Cleanup SymbolSolvent
Melting Point-47°C
CAS108-94-1
ChemAlert Storage SymbolRed
Acidity0.02% Max (as CH3COOH)
View more

Detailed specifications PDF attached below