Potassium Oxalate, Monohydrate (SQ), Qualigens
Qualigens

Potassium Oxalate, Monohydrate (SQ), Qualigens

Have Questions?
Catalog NumberQuantityPackaging
Q19845500 gPlastic Bottles
Catalog number Q19845
HSN 29171190
Price (INR)
1,260.00
Each
Quantity:
500 g
Packaging:
Plastic Bottles
Request bulk or custom format
Price (INR)
1,260.00
Each
Specifications
CAS6487-48-5
ChemAlert Storage SymbolBlue
Melting Point356°C
DOT InformationDOT Class 6.1, : Poison
Density2.13g/cm³
View more

Detailed specifications PDF attached below