Acetic Acid, Glacial, (SQ), Qualigens
Qualigens

Acetic Acid, Glacial, (SQ), Qualigens

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
500 mL
2.5 L
Packaging:
View all
Plastic Drum
Catalog number Q2105C
HSN 29152100
Price (INR)
5,676.00
Each
Quantity:
25 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (INR)
5,676.00
Each
Specifications
CAS64-19-7
Melting Point16°C
Boiling Point117°C
Health Hazard 2DANGER!
Health Hazard 3Emergency Overview
Flammable liquid and vapor. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:2
Instability:0
View more

Detailed specifications PDF attached below