Specifications
Melting Point100°C
CAS10125-13-0
ChemAlert Storage SymbolWhite
Health Hazard 1Signal Word: WARNING!
Health Hazard 3Emergency Overview
Toxic if swallowed. Irritating to eyes, respiratory system and skin. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Corrosive to metal in aqueous
View more